missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-difluorophenol 98.0+%, TCI America™

Catalog No. B44905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44905G 5 g
B449025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44905G Supplier TCI America Supplier No. B44905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 186590-26-1
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD01631350
InChI Key QAHCQGXGAYRHHW-UHFFFAOYSA-N
PubChem CID 2773307
IUPAC Name 5-bromo-2,3-difluorophenol
SMILES C1=C(C=C(C(=C1F)F)O)Br

Specifications

Color Yellow
Quantity 5 g
Synonym 2,3-difluoro-5-bromophenol, phenol, 5-bromo-2,3-difluoro, 5-bromo-2,3-difluoro-phenol, 3-bromo-5,6-difluorophenol, pubchem2843, acmc-209eoh, ksc494o6r, 5-bromo-2,3-difluoro phenol, phenol,5-bromo-2,3-difluoro, 2.3-difluoro-5-bromophenol
Formula Weight 208.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2,3-difluorophenol