missing translation for 'onlineSavingsMsg'
Learn More

5″-Bromo-2,2':5',2″-terthiophene-5-carboxaldehyde 94.0+%, TCI America™

Catalog No. B20581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20581G 1 g
B20585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20581G Supplier TCI America Supplier No. B20581G

Chemical Identifiers

CAS 161726-69-8
Molecular Formula C13H7BrOS3
Molecular Weight (g/mol) 355.28
MDL Number MFCD02093442
InChI Key DSYKMLTVEGWLFR-UHFFFAOYSA-N
PubChem CID 14942412
IUPAC Name 5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O

Specifications

Melting Point 164°C
Quantity 1 g
Synonym 5-Bromo-5′C′C-formyl-2,2′C:5′C,2′C′C-terthiophene
Formula Weight 355.28
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′′-Bromo-2,2′:5′,2′′-terthiophene-5-carboxaldehyde