accessibility menu, dialog, popup

UserName

CAS RN 161726-69-8

IUPAC Name: 5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
Synonyms: 5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
Molecular Weight (g/mol): 355.28
Molecular Formula: C13H7BrOS3
InChi Key: DSYKMLTVEGWLFR-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results