missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-pentanol 90.0+%, TCI America™

Catalog No. p-7119302
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18485G 5 g
B184825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18485G Supplier TCI America Supplier No. B18485G
Only null left

Chemical Identifiers

CAS 34626-51-2
Molecular Formula C5H11BrO
Molecular Weight (g/mol) 167.046
MDL Number MFCD00061101
InChI Key WJVQJXVMLRGNGA-UHFFFAOYSA-N
PubChem CID 118709
IUPAC Name 5-bromopentan-1-ol
SMILES C(CCO)CCBr

Specifications

Boiling Point 117°C
Quantity 5 g
Formula Weight 167.05
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-1-pentanol
EINECSNumber (2)-2002
TSCA Yes
Recommended Storage Freezer