accessibility menu, dialog, popup

UserName

HOBr

CAS RN 34626-51-2

IUPAC Name: 5-bromopentan-1-ol
Synonyms: 5-bromopentan-1-ol
Molecular Weight (g/mol): 167.05
Molecular Formula: C5H11BrO
InChi Key: WJVQJXVMLRGNGA-UHFFFAOYSA-N
SMILES: OCCCCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results