missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AC431090010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC431090010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431090010 Supplier Thermo Scientific Chemicals Supplier No. 431090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 307308-03-8
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
InChI Key CSPINDKIWMCRNX-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydropyrazolo 1,5-a pyridine-3-carboxaldehyde, 4h,5h,6h,7h-pyrazolo 1,5-a pyridine-3-carbaldehyde, 4,5,6,7-tetrahydro-pyrazolo 1,5-a pyridine-3-carbaldehyde, pyrazolo 1,5-a pyridine-3-carboxaldehyde,4,5,6,7-tetrahydro-9ci, pyrazolo 1,5-a pyridine-3-carboxaldehyde, 4,5,6,7-tetrahydro-9ci
PubChem CID 45083680
IUPAC Name 4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde
SMILES C1CCN2C(=C(C=N2)C=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 150.18
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only