accessibility menu, dialog, popup

UserName

CAS RN 307308-03-8

IUPAC Name: 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carbaldehyde
Synonyms: 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carbaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: CSPINDKIWMCRNX-UHFFFAOYSA-N
SMILES: O=CC1=C2CCCCN2N=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results