missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,2,3-trimethoxybenzene 98.0+%, TCI America™

Catalog No. p-7123713
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47395G 5 g
B473925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47395G Supplier TCI America Supplier No. B47395G
Only null left

Chemical Identifiers

CAS 2675-79-8
Molecular Formula C9H11BrO3
Molecular Weight (g/mol) 247.09
MDL Number MFCD00017169
InChI Key XAOOZMATJDXDQJ-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trimethoxybenzene, 3,4,5-trimethoxybromobenzene, bromotrimethoxybenzene, benzene, 5-bromo-1,2,3-trimethoxy, 5-bromo-1,2,3-trimethoxy-benzene, pubchem10980, 3,4,5-trimethoxybromoben, ksc490o1d, 3,4,5-trimethoxyphenylbromide, 3,4,5-triemethoxyphenylbromide
PubChem CID 75885
IUPAC Name 5-bromo-1,2,3-trimethoxybenzene
SMILES COC1=CC(Br)=CC(OC)=C1OC

Specifications

Melting Point 80°C
Boiling Point 273°C
Quantity 5 g
Formula Weight 247.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1,2,3-trimethoxybenzene