missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,2,3-trimethoxybenzene, 97%

Catalog No. p-7027835
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2487206 5 g
AAB2487214 25 g
AAB2487222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2487206 Supplier Thermo Scientific Chemicals Supplier No. B2487206
Only null left

CAS: 2675-79-8 | C9H11BrO3 | 247.09 g/mol

5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 2675-79-8
Molecular Formula C9H11BrO3
Molecular Weight (g/mol) 247.09
MDL Number MFCD00017169
InChI Key XAOOZMATJDXDQJ-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trimethoxybenzene, 3,4,5-trimethoxybromobenzene, bromotrimethoxybenzene, benzene, 5-bromo-1,2,3-trimethoxy, 5-bromo-1,2,3-trimethoxy-benzene, pubchem10980, 3,4,5-trimethoxybromoben, ksc490o1d, 3,4,5-trimethoxyphenylbromide, 3,4,5-triemethoxyphenylbromide
PubChem CID 75885
IUPAC Name 5-bromo-1,2,3-trimethoxybenzene
SMILES COC1=CC(Br)=CC(OC)=C1OC

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 247.1
Percent Purity 97%
Chemical Name or Material 5-Bromo-1,2,3-trimethoxybenzene
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 220-223-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only