missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1,10-phenanthroline 98.0+%, TCI America™
Supplier: TCI America B55671G
Specifications
| 5-Bromo-1,10-phenanthroline | |
| 119°C | |
| C12H7BrN2 | |
| 1 g | |
| BrC1=CC2=CC=CN=C2C2=NC=CC=C12 | |
| 259.11 | |
| 259.11 | |
| Crystalline Powder |
| 40000-20-2 | |
| White-Yellow | |
| MFCD00046903 | |
| GWKGPQCKIBRXGW-UHFFFAOYSA-N | |
| 5-bromo-1,10-phenanthroline | |
| 148335 | |
| ≥98.0% (T) |
Chemical Identifiers
| 40000-20-2 | |
| 259.11 | |
| GWKGPQCKIBRXGW-UHFFFAOYSA-N | |
| 5-bromo-1,10-phenanthroline |
| C12H7BrN2 | |
| MFCD00046903 | |
| 148335 | |
| BrC1=CC2=CC=CN=C2C2=NC=CC=C12 |
Safety and Handling
TSCA : Yes