accessibility menu, dialog, popup

UserName

CAS RN 40000-20-2

BrNN

CAS RN 40000-20-2

IUPAC Name: 5-bromo-1,10-phenanthroline
Synonyms: 5-bromo-1,10-phenanthroline
Molecular Weight (g/mol): 259.11
Molecular Formula: C12H7BrN2
InChi Key: GWKGPQCKIBRXGW-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=CN=C2C2=NC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results