missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1,10-phenanthroline 98.0+%, TCI America™

Catalog No. B5567200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5567200MG 200 mg
B55671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5567200MG Supplier TCI America Supplier No. B5567200MG
Only null left

Chemical Identifiers

CAS 40000-20-2
Molecular Formula C12H7BrN2
Molecular Weight (g/mol) 259.11
MDL Number MFCD00046903
InChI Key GWKGPQCKIBRXGW-UHFFFAOYSA-N
PubChem CID 148335
IUPAC Name 5-bromo-1,10-phenanthroline
SMILES BrC1=CC2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 200 mg
Formula Weight 259.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1,10-phenanthroline