missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxytryptamine hydrochloride, 98%

Catalog No. AC106110010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106110010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106110010 Supplier Thermo Scientific Chemicals Supplier No. 106110010
Only null left

CAS: 52055-23-9 | C17H18N2O·HCl | 302.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52055-23-9
Molecular Formula C17H18N2O·HCl
Molecular Weight (g/mol) 302.8
MDL Number MFCD00012685
InChI Key IUWVJCIEWSQGHH-UHFFFAOYSA-N
Synonym 5-benzyloxytryptamine hydrochloride, 5-benzyloxytryptamine hcl, 2-5-phenylmethoxy-1h-indol-3-yl ethanamine hydrochloride, cambridge id 5119598, c17h18n2o.hcl, 5-benzyloxy-tryptamine hydrochloride, 5-benzyloxy tryptamine hydrochloride, 5-benzyloxytryptamine hydrochloridyl, 3-2-aminoethyl-5-benzyloxyindole hydrochloride
PubChem CID 2828785
IUPAC Name 2-(5-phenylmethoxy-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3CCN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 255°C to 259°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 302.8
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxytryptamine hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 257-626-4

RUO – Research Use Only