accessibility menu, dialog, popup

UserName

CAS RN 52055-23-9

IUPAC Name: 2-[5-(benzyloxy)-1H-indol-3-yl]ethan-1-amine hydrochloridyl
Synonyms: 5-benzyloxytryptamine hydrochloridyl
Molecular Weight (g/mol): 301.79
Molecular Formula: C17H18ClN2O
InChi Key: NIDXFJIFFPKQBY-UHFFFAOYSA-N
SMILES: [Cl].NCCC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results