missing translation for 'onlineSavingsMsg'
Learn More

5-Benzoylpentanoic Acid 98.0+%, TCI America™

Catalog No. p-7119532
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B21825G 5 g
B218225G 25 g
2 options

Catalog No. B21825G

Supplier: TCI America B21825G

Only null left

Chemical Identifiers

CAS 4144-62-1
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00014380
InChI Key AIEMSTCGCMIJTI-UHFFFAOYSA-N
Synonym 5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid
PubChem CID 223595
IUPAC Name 6-oxo-6-phenylhexanoic acid
SMILES OC(=O)CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzoylpentanoic Acid