accessibility menu, dialog, popup

UserName

CAS RN 4144-62-1

IUPAC Name: 6-oxo-6-phenylhexanoic acid
Synonyms: 6-oxo-6-phenylhexanoic acid
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: AIEMSTCGCMIJTI-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)