missing translation for 'onlineSavingsMsg'
Learn More

5-Benzoylpentanoic acid, 99%

Catalog No. aal0829003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0829003 1 g
AAL0829006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0829003 Supplier Thermo Scientific Chemicals Supplier No. L0829003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4144-62-1 | C12H14O3 | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4144-62-1
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00014380
InChI Key AIEMSTCGCMIJTI-UHFFFAOYSA-N
Synonym 5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid
PubChem CID 223595
IUPAC Name 6-oxo-6-phenylhexanoic acid
SMILES OC(=O)CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
Beilstein 1961325
Formula Weight 206.24
Percent Purity 99%
Chemical Name or Material 5-Benzoylpentanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only