missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Benzoylamino)valeric Acid 98.0+%, TCI America™
$55.92 - $505.16
Chemical Identifiers
| CAS | 15647-47-9 |
|---|---|
| Molecular Formula | C12H15NO3 |
| Molecular Weight (g/mol) | 221.256 |
| MDL Number | MFCD00021790 |
| InChI Key | LAPZULVOGQPDBE-UHFFFAOYSA-N |
| PubChem CID | 225176 |
| IUPAC Name | 5-benzamidopentanoic acid |
| SMILES | C1=CC=C(C=C1)C(=O)NCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B0101100MG
|
TCI America
B0101100MG |
100 mg |
N/A
|
|
|||||
|
B01011G
|
TCI America
B01011G |
1 g |
N/A
|
|
|||||
|
B01015G
|
TCI America
B01015G |
5 g |
N/A
|
|
|||||
Chemical Identifiers
| 15647-47-9 | |
| 221.256 | |
| LAPZULVOGQPDBE-UHFFFAOYSA-N | |
| 5-benzamidopentanoic acid |
| C12H15NO3 | |
| MFCD00021790 | |
| 225176 | |
| C1=CC=C(C=C1)C(=O)NCCCCC(=O)O |
Specifications
| 15647-47-9 | |
| White | |
| MFCD00021790 | |
| LAPZULVOGQPDBE-UHFFFAOYSA-N | |
| 5-benzamidopentanoic acid | |
| 225176 | |
| ≥98.0% (T) | |
| 5-(Benzoylamino)valeric Acid |
| 94°C | |
| C12H15NO3 | |
| 100 mg | |
| C1=CC=C(C=C1)C(=O)NCCCCC(=O)O | |
| 221.256 | |
| 221.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No