accessibility menu, dialog, popup

UserName

HOONHO

CAS RN 15647-47-9

IUPAC Name: 5-(phenylformamido)pentanoic acid
Synonyms: 5-(phenylformamido)pentanoic acid
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: LAPZULVOGQPDBE-UHFFFAOYSA-N
SMILES: OC(=O)CCCCNC(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results