missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Benzoylamino)valeric Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 94°C |
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 221.26 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-(Benzoylamino)valeric Acid |
Safety and Handling
| TSCA | No |