missing translation for 'onlineSavingsMsg'
Learn More

5-Azoniaspiro[4.4]nonane Bromide 98.0+%, TCI America™

Catalog No. A30911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30911G 1 g
A30915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30911G Supplier TCI America Supplier No. A30911G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16450-38-7
Molecular Formula C8H16BrN
Molecular Weight (g/mol) 206.127
InChI Key PQTWYBSYKLXZME-UHFFFAOYSA-M
PubChem CID 11458396
IUPAC Name 5-azoniaspiro[4.4]nonane;bromide
SMILES C1CC[N+]2(C1)CCCC2.[Br-]

Specifications

Melting Point 273°C
Color White
Quantity 1 g
Formula Weight 206.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azoniaspiro[4.4]nonane Bromide