accessibility menu, dialog, popup

UserName

CAS RN 16450-38-7

IUPAC Name: 5λ⁵-azaspiro[4.4]nonan-5-ylium bromide
Synonyms: 5λ⁵-azaspiro[4.4]nonan-5-ylium bromide
Molecular Weight (g/mol): 206.13
Molecular Formula: C8H16BrN
InChi Key: PQTWYBSYKLXZME-UHFFFAOYSA-M
SMILES: [Br-].C1CC[N+]2(C1)CCCC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results