missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azaindole, 98%
Description
5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.
Solubility
Hardly soluble in water. Soluble in methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.
Solubility
Hardly soluble in water. Soluble in methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 104°C to 108°C |
| Quantity | 1 g |
| Beilstein | 109684 |
| Solubility Information | Hardly soluble in water. Soluble in methanol and chloroform. |
| Formula Weight | 118.14 |
| Percent Purity | 98% |
| Physical Form | Crystals or Powder or Crystalline Powder |
| Chemical Name or Material | 5-Azaindole |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H StatementH315-H319-H335 Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| RTECSNumber | UY8725000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only