accessibility menu, dialog, popup

UserName

HNN

CAS RN 271-34-1

IUPAC Name: 1H-pyrrolo[3,2-c]pyridine
Synonyms: 5-azaindole
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: SRSKXJVMVSSSHB-UHFFFAOYSA-N
SMILES: N1C=CC2=CN=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results