missing translation for 'onlineSavingsMsg'
Learn More

5-Aminouracil, 97%

CAS: 932-52-5 | C4H5N3O2 | 127.10 g/mol

Supplier:  Thermo Scientific Chemicals L0445214

 View more versions of this product

Catalog No. AAL0445214


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>300°C
932-52-5
MFCD00006025
5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
NC1=CNC(=O)NC1=O
25 g
13611
127.1
5-Aminouracil
C4H5N3O2
127250
BISHACNKZIBDFM-UHFFFAOYSA-N
5-amino-1H-pyrimidine-2,4-dione
127.10
CHEBI:46348
97%

Chemical Identifiers

932-52-5
127.10
BISHACNKZIBDFM-UHFFFAOYSA-N
13611
5-amino-1H-pyrimidine-2,4-dione
C4H5N3O2
MFCD00006025
5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
CHEBI:46348
NC1=CNC(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-252-3

RTECSNumber : YQ8740000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only