missing translation for 'onlineSavingsMsg'
Learn More

5-Aminouracil, 97%

Catalog No. p-7044083
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0445206 5 g
AAL0445214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0445206 Supplier Thermo Scientific Chemicals Supplier No. L0445206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 932-52-5 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-52-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006025
InChI Key BISHACNKZIBDFM-UHFFFAOYSA-N
Synonym 5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
PubChem CID 13611
ChEBI CHEBI:46348
IUPAC Name 5-amino-1H-pyrimidine-2,4-dione
SMILES NC1=CNC(=O)NC1=O

Specifications

Melting Point >300°C
Beilstein 127250
Quantity 5 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 5-Aminouracil
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-252-3
RTECSNumber YQ8740000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only