missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Aminosulfonyl)-2,3-dimethoxybenzoic Acid 98.0+%, TCI America™
$67.38 - $196.36
Chemical Identifiers
| CAS | 66644-80-2 |
|---|---|
| Molecular Formula | C9H11NO6S |
| Molecular Weight (g/mol) | 261.25 |
| MDL Number | MFCD01317561 |
| InChI Key | YEVQOPOKMKTXMD-UHFFFAOYSA-N |
| Synonym | 2,3-Dimethoxy-5-sulfamoylbenzoic Acid |
| PubChem CID | 3017698 |
| IUPAC Name | 2,3-dimethoxy-5-sulfamoylbenzoic acid |
| SMILES | COC1=CC(=CC(C(O)=O)=C1OC)S(N)(=O)=O |
Chemical Identifiers
| 66644-80-2 | |
| 261.25 | |
| YEVQOPOKMKTXMD-UHFFFAOYSA-N | |
| 3017698 | |
| COC1=CC(=CC(C(O)=O)=C1OC)S(N)(=O)=O |
| C9H11NO6S | |
| MFCD01317561 | |
| 2,3-Dimethoxy-5-sulfamoylbenzoic Acid | |
| 2,3-dimethoxy-5-sulfamoylbenzoic acid |
Specifications
| 66644-80-2 | |
| White-Yellow | |
| MFCD01317561 | |
| 2,3-Dimethoxy-5-sulfamoylbenzoic Acid | |
| COC1=CC(=CC(C(O)=O)=C1OC)S(N)(=O)=O | |
| 261.25 | |
| 261.25 | |
| Crystalline Powder |
| 206°C | |
| C9H11NO6S | |
| 5 g | |
| YEVQOPOKMKTXMD-UHFFFAOYSA-N | |
| 2,3-dimethoxy-5-sulfamoylbenzoic acid | |
| 3017698 | |
| ≥98.0% (HPLC,T) | |
| 5-(Aminosulfonyl)-2,3-dimethoxybenzoic Acid |
Safety and Handling
TSCA : No