missing translation for 'onlineSavingsMsg'
Learn More

5-Aminosalicylic Acid 98.0+%, TCI America™

Catalog No. A031725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A031725G 25 g
A0317500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A031725G Supplier TCI America Supplier No. A031725G

Chemical Identifiers

CAS 89-57-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007877
InChI Key KBOPZPXVLCULAV-UHFFFAOYSA-N
Synonym 5-aminosalicylic acid, mesalazine, mesalamine, pentasa, claversal, asacol, canasa, salofalk, rowasa, 5-asa
PubChem CID 4075
ChEBI CHEBI:6775
IUPAC Name 5-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)C(=O)O)O

Specifications

Melting Point 265°C
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminosalicylic Acid
EINECSNumber (3)-1581
RTECSNumber VO1400000
TSCA Yes