missing translation for 'onlineSavingsMsg'
Learn More

5-Aminosalicylic acid, 99%

Catalog No. AC134330050
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC134330050 5 g
AC134330250 25 g
AC134331000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AC134330050 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 134330050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89-57-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-57-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007877
InChI Key KBOPZPXVLCULAV-UHFFFAOYSA-N
Synonym 5-aminosalicylic acid, mesalazine, mesalamine, pentasa, claversal, asacol, canasa, salofalk, rowasa, 5-asa
PubChem CID 4075
ChEBI CHEBI:6775
IUPAC Name 5-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1N)C(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 283.0°C
Color Beige-Brown to Gray
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(OH)CO2H
Quantity 5 g
Beilstein 14, 579
Merck Index 15, 5974
Solubility Information Solubility in water: 0.84g/L (20°C). Other solubilities: better soluble in hot water, soluble in hcl and diluted naoh, insoluble in ethanol
Formula Weight 153.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Aminosalicylic acid, 99%
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-919-1
RTECSNumber VO1400000
TSCA TSCA

RUO – Research Use Only