Learn More
5-Aminoquinoline, 99%
CAS: 611-34-7 | C9H8N2 | 144.18 g/mol
Supplier: Thermo Scientific Chemicals L1962806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Aminoquinoline | |
| 107°C to 109°C | |
| C9H8N2 | |
| 5 g | |
| Air Sensitive | |
| XMIAFAKRAAMSGX-UHFFFAOYSA-N | |
| quinolin-5-amine | |
| 11911 | |
| 99% |
| 611-34-7 | |
| 310°C | |
| MFCD00006797 | |
| 114479 | |
| 5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh | |
| NC1=C2C=CC=NC2=CC=C1 | |
| 144.18 | |
| 144.18 |
Chemical Identifiers
| 611-34-7 | |
| 144.18 | |
| XMIAFAKRAAMSGX-UHFFFAOYSA-N | |
| 11911 | |
| NC1=C2C=CC=NC2=CC=C1 |
| C9H8N2 | |
| MFCD00006797 | |
| 5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh | |
| quinolin-5-amine |
Safety and Handling
GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 210-266-1
RTECSNumber : VA9625000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only