missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoquinoline, 99%

Catalog No. p-7047686
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962803 1 g
AAL1962806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1962803 Supplier Thermo Scientific Chemicals Supplier No. L1962803
Only null left

CAS: 611-34-7 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-34-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006797
InChI Key XMIAFAKRAAMSGX-UHFFFAOYSA-N
Synonym 5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh
PubChem CID 11911
IUPAC Name quinolin-5-amine
SMILES NC1=C2C=CC=NC2=CC=C1

Specifications

Melting Point 107°C to 109°C
Boiling Point 310°C
Quantity 1 g
Beilstein 114479
Sensitivity Air Sensitive
Formula Weight 144.18
Percent Purity 99%
Chemical Name or Material 5-Aminoquinoline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-266-1
RTECSNumber VA9625000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only