missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoquinoline, 99%

CAS: 611-34-7 | C9H8N2 | 144.18 g/mol

Supplier:  Thermo Scientific Chemicals L1962803

 View more versions of this product

Catalog No. AAL1962803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Aminoquinoline
107°C to 109°C
C9H8N2
1 g
Air Sensitive
XMIAFAKRAAMSGX-UHFFFAOYSA-N
quinolin-5-amine
11911
99%
611-34-7
310°C
MFCD00006797
114479
5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh
NC1=C2C=CC=NC2=CC=C1
144.18
144.18

Chemical Identifiers

611-34-7
144.18
XMIAFAKRAAMSGX-UHFFFAOYSA-N
11911
NC1=C2C=CC=NC2=CC=C1
C9H8N2
MFCD00006797
5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh
quinolin-5-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 210-266-1

RTECSNumber : VA9625000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only