missing translation for 'onlineSavingsMsg'
Learn More

5-Aminopyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. A23861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23861G 1 g
A23865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23861G Supplier TCI America Supplier No. A23861G

Chemical Identifiers

CAS 24242-20-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD02684600
InChI Key WDJARUKOMOGTHA-UHFFFAOYSA-N
Synonym 5-amino-2-pyridinecarboxylic acid, 5-aminopicolinic acid, 5-amino-2-picolinic acid, 2-pyridinecarboxylic acid, 5-amino, 5-amino-pyridine-2-carboxylic acid, 5-amino-2-pyridinecarboxylicacid, pubchem15896, 5-amino-2-carboxypyridine, acmc-209ga3, ksc201o9n
PubChem CID 4179726
IUPAC Name 5-aminopyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1N)C(=O)O

Specifications

Melting Point 218°C
Quantity 1 g
Formula Weight 138.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminopyridine-2-carboxylic Acid