missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonicotinic Acid 98.0+%, TCI America™

Catalog No. p-7117679
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20005G 5 g
A200025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20005G Supplier TCI America Supplier No. A20005G
Only null left

Chemical Identifiers

CAS 24242-19-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00129116
InChI Key BYIORJAACCWFPU-UHFFFAOYSA-N
Synonym 5-aminonicotinic acid, 5-amino-nicotinic acid, 5-aminonicotinicacid, 3-pyridinecarboxylic acid, 5-amino, 5-amino nicotinic acid, 5-amino-3-carboxypyridine, 5-amino-3-pyridinecarboxylic acid, zlchem 307, pubchem15914, acmc-209ga2
PubChem CID 354316
ChEBI CHEBI:68578
IUPAC Name 5-aminopyridine-3-carboxylic acid
SMILES C1=C(C=NC=C1N)C(=O)O

Specifications

Melting Point 293°C
Color Yellow
Quantity 5 g
Formula Weight 138.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminonicotinic Acid