accessibility menu, dialog, popup

UserName

NH2NOO

CAS RN 24242-19-1

IUPAC Name: 5-aminopyridine-3-carboxylate
Synonyms: 5-aminopyridine-3-carboxylate
Molecular Weight (g/mol): 137.12
Molecular Formula: C6H5N2O2
InChi Key: BYIORJAACCWFPU-UHFFFAOYSA-M
SMILES: NC1=CN=CC(=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)