Learn More
5-Aminonaphthalene-2-sulfonic acid, 97%
CAS: 119-79-9 | C10H9NO3S | 223.246 g/mol
Supplier: Thermo Scientific Chemicals L0309914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Aminonaphthalene-2-sulfonic acid | |
| >300°C | |
| C10H9NO3S | |
| 25 g | |
| 14,2350 | |
| UWPJYQYRSWYIGZ-UHFFFAOYSA-N | |
| 5-aminonaphthalene-2-sulfonic acid | |
| 8408 | |
| 223.25 |
| 119-79-9 | |
| Odorless | |
| MFCD00004030 | |
| 1819887 | |
| 5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid | |
| C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N | |
| 223.246 | |
| CHEBI:44188 | |
| 97% |
Chemical Identifiers
| 119-79-9 | |
| 223.246 | |
| UWPJYQYRSWYIGZ-UHFFFAOYSA-N | |
| 8408 | |
| 5-aminonaphthalene-2-sulfonic acid |
| C10H9NO3S | |
| MFCD00004030 | |
| 5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid | |
| CHEBI:44188 | |
| C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N |
Safety and Handling
EINECSNumber : 204-351-2
RTECSNumber : QK1285000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only