missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoisoquinoline 98.0+%, TCI America™
$29.83 - $89.48
Chemical Identifiers
| CAS | 1125-60-6 |
|---|---|
| Molecular Formula | C9H8N2 |
| Molecular Weight (g/mol) | 144.18 |
| MDL Number | MFCD00006907 |
| InChI Key | DTVYNUOOZIKEEX-UHFFFAOYSA-N |
| Synonym | 5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline |
| PubChem CID | 70766 |
| IUPAC Name | isoquinolin-5-amine |
| SMILES | NC1=C2C=CN=CC2=CC=C1 |
Chemical Identifiers
| 1125-60-6 | |
| 144.18 | |
| DTVYNUOOZIKEEX-UHFFFAOYSA-N | |
| 70766 | |
| NC1=C2C=CN=CC2=CC=C1 |
| C9H8N2 | |
| MFCD00006907 | |
| 5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline | |
| isoquinolin-5-amine |
Specifications
| 1125-60-6 | |
| C9H8N2 | |
| 1 g | |
| DTVYNUOOZIKEEX-UHFFFAOYSA-N | |
| isoquinolin-5-amine | |
| 70766 | |
| ≥98.0% (GC,T) | |
| 5-Aminoisoquinoline |
| 130°C | |
| MFCD00006907 | |
| 5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline | |
| NC1=C2C=CN=CC2=CC=C1 | |
| 144.18 | |
| 144.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No