accessibility menu, dialog, popup

UserName

CAS RN 1125-60-6

IUPAC Name: isoquinolin-5-amine
Synonyms: 5-aminoisoquinoline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: DTVYNUOOZIKEEX-UHFFFAOYSA-N
SMILES: NC1=C2C=CN=CC2=CC=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (4)