missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoisoquinoline 98.0+%, TCI America™

Catalog No. A14651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14651G 1 g
A14655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14651G Supplier TCI America Supplier No. A14651G

Chemical Identifiers

CAS 1125-60-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006907
InChI Key DTVYNUOOZIKEEX-UHFFFAOYSA-N
Synonym 5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline
PubChem CID 70766
IUPAC Name isoquinolin-5-amine
SMILES NC1=C2C=CN=CC2=CC=C1

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoisoquinoline