missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoisoquinoline, 99%

Catalog No. AAL0122303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0122303 1 g
AAL0122306 5 g
AAL0122314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0122303 Supplier Thermo Scientific Chemicals Supplier No. L0122303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1125-60-6 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1125-60-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006907
InChI Key DTVYNUOOZIKEEX-UHFFFAOYSA-N
Synonym 5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline
PubChem CID 70766
IUPAC Name isoquinolin-5-amine
SMILES NC1=C2C=CN=CC2=CC=C1

Specifications

Melting Point 126°C to 131°C
Quantity 1 g
Beilstein 114465
Sensitivity Light sensitive
Formula Weight 144.18
Percent Purity 99%
Chemical Name or Material 5-Aminoisoquinoline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-408-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only