missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoisophthalic Acid Hydrate 98.0+%, TCI America™
Supplier: TCI America A1290100G
Specifications
| 5-Aminoisophthalic Acid Hydrate | |
| White-Yellow | |
| MFCD00002522 | |
| 5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous | |
| C1=C(C=C(C=C1C(=O)O)N)C(=O)O | |
| 181.147 | |
| 181.15 | |
| Crystalline Powder |
| 99-31-0 | |
| C8H7NO4 | |
| 100 g | |
| KBZFDRWPMZESDI-UHFFFAOYSA-N | |
| 5-aminobenzene-1,3-dicarboxylic acid | |
| 66833 | |
| ≥98.0% (T) |
Chemical Identifiers
| 99-31-0 | |
| 181.147 | |
| KBZFDRWPMZESDI-UHFFFAOYSA-N | |
| 66833 | |
| C1=C(C=C(C=C1C(=O)O)N)C(=O)O |
| C8H7NO4 | |
| MFCD00002522 | |
| 5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous | |
| 5-aminobenzene-1,3-dicarboxylic acid |
Safety and Handling
TSCA : Yes