accessibility menu, dialog, popup

UserName

CAS RN 99-31-0

IUPAC Name: 5-aminobenzene-1,3-dicarboxylic acid
Synonyms: 5-aminobenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: KBZFDRWPMZESDI-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)