missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoisophthalic Acid Hydrate 98.0+%, TCI America™

Catalog No. A1290500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1290500G 500 g
A129025G 25 g
A1290100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1290500G Supplier TCI America Supplier No. A1290500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-31-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00002522
InChI Key KBZFDRWPMZESDI-UHFFFAOYSA-N
Synonym 5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
PubChem CID 66833
IUPAC Name 5-aminobenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)N)C(=O)O

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoisophthalic Acid Hydrate