missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindane, 97%

Catalog No. AAL0250409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0250409 10 g
AAL0250418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0250409 Supplier Thermo Scientific Chemicals Supplier No. L0250409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24425-40-9 | C9H11N | 133.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24425-40-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD00003803
InChI Key LEWZOBYWGWKNCK-UHFFFAOYSA-N
Synonym 5-aminoindan, indan-5-amine, indan-5-ylamine, 5-indanamine, 5-aminohydrindene, 5-aminoindane, indane-5-ylamine, 1h-inden-5-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-5-ylamine, 6-aminoindane
PubChem CID 90496
IUPAC Name 2,3-dihydro-1H-inden-5-amine
SMILES C1CC2=C(C1)C=C(C=C2)N

Specifications

Melting Point 32°C to 37°C
Boiling Point 248°C to 250°C
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN2811
Beilstein 2082278
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material 5-Aminoindane
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 246-241-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only