missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindan 98.0+%, TCI America™

Catalog No. A099110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A099110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A099110G Supplier TCI America Supplier No. A099110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24425-40-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD00003803
InChI Key LEWZOBYWGWKNCK-UHFFFAOYSA-N
Synonym 5-aminoindan, indan-5-amine, indan-5-ylamine, 5-indanamine, 5-aminohydrindene, 5-aminoindane, indane-5-ylamine, 1h-inden-5-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-5-ylamine, 6-aminoindane
PubChem CID 90496
IUPAC Name 2,3-dihydro-1H-inden-5-amine
SMILES C1CC2=C(C1)C=C(C=C2)N

Specifications

Melting Point 37°C
Boiling Point 147°C
Quantity 10 g
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoindan
TSCA No
Recommended Storage Refrigerator