missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindan, 97%

Catalog No. AC157350500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157350500 50 g Glass bottle
AC157350100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157350500 Supplier Thermo Scientific Chemicals Supplier No. 157350500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24425-40-9 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24425-40-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00003803
InChI Key LEWZOBYWGWKNCK-UHFFFAOYSA-N
Synonym 5-aminoindan, indan-5-amine, indan-5-ylamine, 5-indanamine, 5-aminohydrindene, 5-aminoindane, indane-5-ylamine, 1h-inden-5-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-5-ylamine, 6-aminoindane
PubChem CID 90496
IUPAC Name 2,3-dihydro-1H-inden-5-amine
SMILES C1CC2=C(C1)C=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34.0°C to 36.0°C
Color Brown
Boiling Point 247.0°C to 249.0°C (745.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 50 g
Beilstein 12, I, 511
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether, acetone, benzene
Formula Weight 133.19
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 5-Aminoindan, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 246-241-7

RUO – Research Use Only