missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-8-hydroxyquinoline Dihydrochloride 98.0+%, TCI America™

Catalog No. A100525G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A100525G 25 g
A10055G 5 g
2 options

Catalog No. A100525G

Supplier: TCI America A100525G

Only null left

Chemical Identifiers

CAS 21302-43-2
Molecular Formula C9H10Cl2N2O
Molecular Weight (g/mol) 233.092
MDL Number MFCD00012737
InChI Key VTQDJAUGGZFPOI-UHFFFAOYSA-N
Synonym 5-Amino-8-quinolinol Dihydrochloride
PubChem CID 16211947
IUPAC Name 5-aminoquinolin-8-ol;dihydrochloride
SMILES C1=CC2=C(C=CC(=C2N=C1)O)N.Cl.Cl

Specifications

Quantity 25 g
Formula Weight 233.09
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-8-hydroxyquinoline Dihydrochloride