accessibility menu, dialog, popup

UserName

CAS RN 21302-43-2

IUPAC Name: 5-aminoquinolin-8-ol hydrochloridyl
Synonyms: 8-quinolinol, 5-amino- hydrochloridyl
Molecular Weight (g/mol): 195.63
Molecular Formula: C9H8ClN2O
InChi Key: CPWUQKCQXWSDMF-UHFFFAOYSA-N
SMILES: [Cl].NC1=C2C=CC=NC2=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results