missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-4,6-dichloropyrimidine 98.0+%, TCI America™
$41.02 - $93.21
Chemical Identifiers
| CAS | 5413-85-4 |
|---|---|
| Molecular Formula | C4H3Cl2N3 |
| Molecular Weight (g/mol) | 163.99 |
| MDL Number | MFCD00006108 |
| InChI Key | NIGDWBHWHVHOAD-UHFFFAOYSA-N |
| Synonym | 5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine |
| PubChem CID | 79434 |
| IUPAC Name | 4,6-dichloropyrimidin-5-amine |
| SMILES | NC1=C(Cl)N=CN=C1Cl |
Chemical Identifiers
| 5413-85-4 | |
| 163.99 | |
| NIGDWBHWHVHOAD-UHFFFAOYSA-N | |
| 79434 | |
| NC1=C(Cl)N=CN=C1Cl |
| C4H3Cl2N3 | |
| MFCD00006108 | |
| 5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine | |
| 4,6-dichloropyrimidin-5-amine |
Specifications
| 5413-85-4 | |
| White-Yellow | |
| MFCD00006108 | |
| 5-amino-4,6-dichloropyrimidine, 5-pyrimidinamine, 4,6-dichloro, 4,6-dichloro-5-pyrimidinamine, 4,6-dichloro-5-aminopyrimidine, 4,6-dichloropyrimidin-5-ylamine, 5-amine-4,6-dichloropyrimidine, 5-amino-4,6-dichloropyfimidine, 4,6-dichloro-pyrimidin-5-ylamine, pyrimidine, 5-amino-4,6-dichloro, 4,6-dichloropyrimidine-5-ylamine | |
| NC1=C(Cl)N=CN=C1Cl | |
| 163.99 | |
| 163.99 | |
| Crystalline Powder |
| 144°C | |
| C4H3Cl2N3 | |
| 1 g | |
| NIGDWBHWHVHOAD-UHFFFAOYSA-N | |
| 4,6-dichloropyrimidin-5-amine | |
| 79434 | |
| ≥98.0% (N) | |
| 5-Amino-4,6-dichloropyrimidine |
Safety and Handling
TSCA : Yes