accessibility menu, dialog, popup

UserName

CAS RN 5413-85-4

IUPAC Name: 4,6-dichloropyrimidin-5-amine
Synonyms: 4,6-dichloropyrimidin-5-amine
Molecular Weight (g/mol): 163.99
Molecular Formula: C4H3Cl2N3
InChi Key: NIGDWBHWHVHOAD-UHFFFAOYSA-N
SMILES: NC1=C(Cl)N=CN=C1Cl
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)